missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-3'-methoxyflavone, 97%

$62.38 - $560.97

Chemical Identifiers

CAS 76666-32-5
Molecular Formula C16H12O4
Formula Weight 268.26
MDL Number MFCD00017730
Chemical Name or Material 3-Hydroxy-3'-methoxyflavone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AAH2694303 SDS Alfa Aesar™
H2694303
1g Each for $62.38
AAH2694306 SDS Alfa Aesar™
H2694306
5g Each for $185.50
AAH2694314 SDS Alfa Aesar™
H2694314
25g Each for $560.97
Specifications Description & Specifications

Chemical Identifiers

76666-32-5
268.26
3-Hydroxy-3'-methoxyflavone
C16H12O4
MFCD00017730

Specifications

1g
C16H12O4
MFCD00017730
97%
76666-32-5
268.26
3-Hydroxy-3'-methoxyflavone

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning