missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, 97+% (total alcohols), ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 121970250

 View more versions of this product

Catalog No. AC121970250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene
96°C
Glass bottle
0.87
Yellow
25g
7212-44-4
MFCD00008911
--nerolidol, 1,6,10-dodecatrien-3-ol, 3,7,11-trimethyl-, 3r,6e, 3r,6e-3,7,11-trimethyldodeca-1,6,10-trien-3-ol, 3r,6e-nerolidol, 3r-6e-nerolidol, ?-nerolidol, e-nerolidol, nerolidol, nerolidol, 6e--, unii-uoc0644v25
FQTLCLSUCSAZDY-GOFCXVBSSA-N
(3R,6E)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
11241545
222.37
97+% (total alcohols)
0.8700g/mL
Authentic
1.4780 to 1.4830
114.0°C (1.0 mmHg)
-75.0°C
≥97% (total alcohols)
C15H26O
15, 6561
Solubility in water: immiscible. Other solubilities: soluble in abs. alc. and fixed oils, (still clearly sol. in 3 parts of 70% alc.), insoluble in glycerol
CC(=CCCC(=CCCC(C)(C=C)O)C)C
222.37
CHEBI:59959
Liquid

Chemical Identifiers

7212-44-4
222.37
FQTLCLSUCSAZDY-GOFCXVBSSA-N
11241545
(3R,6E)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
C15H26O
MFCD00008911
--nerolidol, 1,6,10-dodecatrien-3-ol, 3,7,11-trimethyl-, 3r,6e, 3r,6e-3,7,11-trimethyldodeca-1,6,10-trien-3-ol, 3r,6e-nerolidol, 3r-6e-nerolidol, ?-nerolidol, e-nerolidol, nerolidol, nerolidol, 6e--, unii-uoc0644v25
CHEBI:59959
CC(=CCCC(=CCCC(C)(C=C)O)C)C
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 230-597-5

SDS Documents