Please login to your online account to display your discounted pricing

3-Hydroxy-2-quinoxalinecarboxylic Acid 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  148590010

 View more versions of this product

Catalog No. AC148590010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder and Chunks
Formula Weight 190.16
Quantity 1g
Color Green-yellow
EINECS Number 214-877-4
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 3-Hydroxy-2-quinoxalinecarboxylic acid
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H6N2O3
MDL Number MFCD00006726
CAS 1204-75-7
CAS Min % 97.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Hydroxy-2-quinoxalinecarboxylic acid
Name Note 98%
CAS 1204-75-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H6N2O3
Formula Weight 190.16
MDL Number MFCD00006726