missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-2-quinoxalinecarboxylic Acid 98%, ACROS Organics™

Chemical Identifiers

CAS 1204-75-7
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C9H6N2O3
Formula Weight 190.16
MDL Number MFCD00006726
Chemical Name or Material 3-Hydroxy-2-quinoxalinecarboxylic acid, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC148590010 SDS Thermo Fisher Scientific
148590010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC148590050 SDS Fisher Scientific Acros
148590050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1204-75-7
100.0
C9H6N2O3
MFCD00006726
≥97.5%
97.5
98%
190.16
3-Hydroxy-2-quinoxalinecarboxylic acid, 98%

Specifications

190.16
1g
100.0
Glass bottle
C9H6N2O3
98%
3-Hydroxy-2-quinoxalinecarboxylic acid, 98%
Powder and Chunks
Green-yellow
97.5
Authentic
MFCD00006726
1204-75-7
≥97.5%

Safety and Handling

EINECSNumber : 214-877-4

TSCA : TSCA