Please login to your online account to display your discounted pricing

3-Hydroxy-2-naphthoic Acid Hydrazide 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  121620250

 View more versions of this product

Catalog No. AC121620250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 202.21
Quantity 25g
Color Beige
Beilstein 10, 336
EINECS Number 226-282-7
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Synonym 3-Hydroxy-2-naphthydrazide
Infrared Spectrum Authentic
Chemical Name or Material 3-Hydroxy-2-naphthoic acid hydrazide
Name Note 97%
Assay Percent Range 97%
Molecular Formula C11H10N2O2
Linear Formula HOC10H6CONHNH2
MDL Number MFCD00004097
CAS 5341-58-2

Specifications

Chemical Name or Material 3-Hydroxy-2-naphthoic acid hydrazide
Name Note 97%
CAS 5341-58-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H10N2O2
Linear Formula HOC10H6CONHNH2
Formula Weight 202.21
MDL Number MFCD00004097
Synonym 3-Hydroxy-2-naphthydrazide