Please login to your online account to display your discounted pricing

3-Hydroxy-2-naphthoic Acid Hydrazide 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-2-naphthoic acid hydrazide
Name Note 97%
CAS 5341-58-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H10N2O2
Linear Formula HOC10H6CONHNH2
Formula Weight 202.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC121620250

  SDS
thermo fisher scientific
121620250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC121620050

  SDS
fisher scientific acros
121620050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 202.21
Quantity 25g
Color Beige
Beilstein 10, 336
TSCA TSCA
EINECS Number 226-282-7
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Synonym 3-Hydroxy-2-naphthydrazide
Infrared Spectrum Authentic
Name Note 97%
Linear Formula HOC10H6CONHNH2
CAS 5341-58-2
Molecular Formula C11H10N2O2
Assay Percent Range 97%
Chemical Name or Material 3-Hydroxy-2-naphthoic acid hydrazide
MDL Number MFCD00004097