missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-2-naphthoic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 121610010

 View more versions of this product

Catalog No. AC121610010



Description

Description

Specifications

Specifications

3-Hydroxy-2-naphthoic acid
Authentic
Beige-Brown to Yellow
1kg
92-70-6
100.0
C11H8O3
10, 333
2-hydroxy-3-naphthoic acid, 2-naphthalenecarboxylic acid, 3-hydroxy, 2-naphthol-3-carboxylic acid, 3-hydroxy-2-naphthoic acid, b.o.n. acid, bon acid, bona, c.i. developer 20, developer bon, miketazol developer ons
ALKYHXVLJMQRLQ-UHFFFAOYSA-N
3-hydroxynaphthalene-2-carboxylic acid
7104
188.18
98%
>150°C
Plastic bottle
217°C to 223°C
98%
97.5
98%
MFCD00004103
15, 4874
Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alkalies,freely soluble in alcohol,ether and benzene
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)O
188.18
CHEBI:80383
Fine Crystalline Powder

Chemical Identifiers

92-70-6
188.18
ALKYHXVLJMQRLQ-UHFFFAOYSA-N
7104
3-hydroxynaphthalene-2-carboxylic acid
C11H8O3
MFCD00004103
2-hydroxy-3-naphthoic acid, 2-naphthalenecarboxylic acid, 3-hydroxy, 2-naphthol-3-carboxylic acid, 3-hydroxy-2-naphthoic acid, b.o.n. acid, bon acid, bona, c.i. developer 20, developer bon, miketazol developer ons
CHEBI:80383
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-180-8

RTECSNumber : QL1755000

TSCA : TSCA

SDS Documents