missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-2-methylpyridine, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 345220050

 View more versions of this product

Catalog No. AC345220050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Hydroxy-2-methylpyridine
98.5
Authentic
Glass bottle
5g
AQSRRZGQRFFFGS-UHFFFAOYSA-N
2-methylpyridin-3-ol
70719
99%
1121-25-1
100.0
98.5% min. (GC)
C6H7NO
3-hydroxy-2-methylpyridine, 2-methyl-3-hydroxypyridine, 3-hydroxy-2-picoline, 2-methyl-3-pyridinol, 3-pyridinol, 2-methyl, 2-methyl-3-hydroxylpyridine, unii-365fj23a8z, 3-pyridinol, methyl, 2-methyl-pyridin-3-ol, 3-hydroxy-2-methylpyridine 3-hydroxy-2-picoline
CC1=C(C=CC=N1)O
109.13
109.13

Chemical Identifiers

1121-25-1
109.13
3-hydroxy-2-methylpyridine, 2-methyl-3-hydroxypyridine, 3-hydroxy-2-picoline, 2-methyl-3-pyridinol, 3-pyridinol, 2-methyl, 2-methyl-3-hydroxylpyridine, unii-365fj23a8z, 3-pyridinol, methyl, 2-methyl-pyridin-3-ol, 3-hydroxy-2-methylpyridine 3-hydroxy-2-picoline
2-methylpyridin-3-ol
C6H7NO
AQSRRZGQRFFFGS-UHFFFAOYSA-N
70719
CC1=C(C=CC=N1)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 214-327-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only