Please login to your online account to display your discounted pricing

3-Hydroxy-2-methylpyridine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-2-methylpyridine
CAS 1121-25-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H7NO
Formula Weight 109.13
Synonym 2-Methyl-3-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC345220050

  SDS
fisher scientific acros
34522-0050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC345220250

  SDS
thermo fisher scientific
AC345220250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder
Formula Weight 109.13
Quantity 5g
Color Beige-brown
Melting Point 167° to 171°C
CAS Max % 100.0
Chemical Name or Material 3-Hydroxy-2-methylpyridine
Assay Percent Range 99%
Molecular Formula C6H7NO
CAS 1121-25-1
Synonym 2-Methyl-3-pyridinol
CAS Min % 98.5
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Safety and Handling

Danger

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.