missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-2-iodopyridine, 98%

$79.55 - $838.96

Chemical Identifiers

CAS 40263-57-8
Molecular Formula C5H4INO
Molecular Weight (g/mol) 220.997
MDL Number MFCD00023421
InChI Key HJBGMPCMSWJZNH-UHFFFAOYSA-N
Synonym 2-iodo-3-hydroxy-pyridine, 2-iodo-3-hydroxypyridin, 2-iodo-3-hydroxypyridine, 2-iodo-3-pyridinol, 2-iodo-pyridin-3-ol, 2-iodopyridine-3-ol, 2-iodpyridin-3-ol, 3-hydroxy-2-iodopyridine, 3-pyridinol, 2-iodo, acmc-209jcf
PubChem CID 97179
IUPAC Name 2-iodopyridin-3-ol
SMILES C1=CC(=C(N=C1)I)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0866703
View Documents
Alfa Aesar™
L0866703
1g
Each for $79.55
 
AAL0866706
View Documents
Alfa Aesar™
L0866706
5g
Each for $237.86
 
AAL0866714
View Documents
Alfa Aesar™
L0866714
25g
Each for $838.96
 
Description

Description

Applications
It is used in the synthesis of isatogens via tolan (diphenylacetylene) intermediates.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
Specifications

Chemical Identifiers

40263-57-8
220.997
HJBGMPCMSWJZNH-UHFFFAOYSA-N
97179
C1=CC(=C(N=C1)I)O
C5H4INO
MFCD00023421
2-iodo-3-hydroxy-pyridine, 2-iodo-3-hydroxypyridin, 2-iodo-3-hydroxypyridine, 2-iodo-3-pyridinol, 2-iodo-pyridin-3-ol, 2-iodopyridine-3-ol, 2-iodpyridin-3-ol, 3-hydroxy-2-iodopyridine, 3-pyridinol, 2-iodo, acmc-209jcf
2-iodopyridin-3-ol

Specifications

Light sensitive
1g
C5H4INO
109836
Slightly soluble in water.
C1=CC(=C(N=C1)I)O
220.997
221
3-Hydroxy-2-iodopyridine
191°C to 195°C
40263-57-8
MFCD00023421
2-iodo-3-hydroxy-pyridine, 2-iodo-3-hydroxypyridin, 2-iodo-3-hydroxypyridine, 2-iodo-3-pyridinol, 2-iodo-pyridin-3-ol, 2-iodopyridine-3-ol, 2-iodpyridin-3-ol, 3-hydroxy-2-iodopyridine, 3-pyridinol, 2-iodo, acmc-209jcf
HJBGMPCMSWJZNH-UHFFFAOYSA-N
2-iodopyridin-3-ol
97179
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 254-864-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS