missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-2-iodopyridine, 98%

Manufacturer:  Alfa Aesar™ L0866703

 View more versions of this product

Catalog No. AAL0866703



Description

Description

Applications
It is used in the synthesis of isatogens via tolan (diphenylacetylene) intermediates.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
Specifications

Specifications

3-Hydroxy-2-iodopyridine
191°C to 195°C
40263-57-8
MFCD00023421
2-iodo-3-hydroxy-pyridine
HJBGMPCMSWJZNH-UHFFFAOYSA-N
2-iodopyridin-3-ol
97179
98%
Light sensitive
1g
C5H4INO
109836
Slightly soluble in water.
C1=CC(=C(N=C1)I)O
220.997
221

Chemical Identifiers

40263-57-8
220.997
HJBGMPCMSWJZNH-UHFFFAOYSA-N
97179
C1=CC(=C(N=C1)I)O
C5H4INO
MFCD00023421
2-iodo-3-hydroxy-pyridine
2-iodopyridin-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 254-864-0

TSCA : No

Recommended Storage : Ambient temperatures

Documents