Learn More
3-Hydroxy-2,4,6-tribromobenzoic acid, 97%, Thermo Scientific Chemicals
$102.69 - $102.69
Chemical Identifiers
CAS | 14348-40-4 |
---|---|
Molecular Formula | C7H3Br3O3 |
Molecular Weight (g/mol) | 374.84 |
MDL Number | MFCD00055557 |
InChI Key | YDBHVMTTYXWHLI-UHFFFAOYSA-N |
Synonym | 3-hydroxy-2,4,6-tribromobenzoic acid, tbhba, benzoic acid, 2,4,6-tribromo-3-hydroxy, 2,4,6-tribromo-3-hydroxybenzoicacid, 2,4,6-thba, pubchem12541, acmc-209uep, ksc174o3h, 2,4,6-tribromo-3-hydroxy-benzoic acid, 2,4,6-tribromo-3-hydroxybenzoic acid |
PubChem CID | 151915 |
IUPAC Name | 2,4,6-tribromo-3-hydroxybenzoic acid |
SMILES | C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC334890050
|
Thermo Scientific Chemicals
334890050 |
5 g | Glass bottle |
Each for $102.69
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14348-40-4 | |
374.84 | |
YDBHVMTTYXWHLI-UHFFFAOYSA-N | |
151915 | |
C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br |
C7H3Br3O3 | |
MFCD00055557 | |
3-hydroxy-2,4,6-tribromobenzoic acid, tbhba, benzoic acid, 2,4,6-tribromo-3-hydroxy, 2,4,6-tribromo-3-hydroxybenzoicacid, 2,4,6-thba, pubchem12541, acmc-209uep, ksc174o3h, 2,4,6-tribromo-3-hydroxy-benzoic acid, 2,4,6-tribromo-3-hydroxybenzoic acid | |
2,4,6-tribromo-3-hydroxybenzoic acid |
Specifications
14348-40-4 | |
100.0 | |
White | |
97% | |
C7H3Br3O3 | |
MFCD00055557 | |
3-hydroxy-2,4,6-tribromobenzoic acid, tbhba, benzoic acid, 2,4,6-tribromo-3-hydroxy, 2,4,6-tribromo-3-hydroxybenzoicacid, 2,4,6-thba, pubchem12541, acmc-209uep, ksc174o3h, 2,4,6-tribromo-3-hydroxy-benzoic acid, 2,4,6-tribromo-3-hydroxybenzoic acid | |
YDBHVMTTYXWHLI-UHFFFAOYSA-N | |
2,4,6-tribromo-3-hydroxybenzoic acid | |
151915 | |
97% | |
3-Hydroxy-2, 4, 6-tribromobenzoic acid, 97% |
96.0 | |
144°C to 149°C | |
Authentic | |
Glass bottle | |
C6HBr3(OH)COOH | |
5 g | |
Solubility in water: insoluble | |
C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br | |
374.84 | |
374.84 | |
Crystals |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA
RUO – Research Use Only