Please login to your online account to display your discounted pricing

3-Hydroxy-2,4,5-trifluorobenzoic acid, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC357300250

 View more versions of this product

Catalog No. AC357300250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 192.09
Quantity 25g
Color White to light yellow-beige
Melting Point 143° to 147°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Linear Formula F3C6H(OH)CO2H
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Assay Percent Range 98%
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00800378
Molecular Formula C7 H3 F3 O3
Chemical Name or Material 3-Hydroxy-2, 4, 5-trifluorobenzoic acid
CAS 116751-24-7

Specifications

Chemical Name or Material 3-Hydroxy-2, 4, 5-trifluorobenzoic acid
Name Note 98%
CAS 116751-24-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H3 F3 O3
Linear Formula F3C6H(OH)CO2H
Formula Weight 192.09
MDL Number MFCD00800378
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents