missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-2,4,5-trifluorobenzoic acid, 98%, ACROS Organics™

$784.38 - $784.38

Chemical Identifiers

CAS 116751-24-7
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7 H3 F3 O3
Linear Formula F3C6H(OH)CO2H
Formula Weight 192.09
MDL Number MFCD00800378
Chemical Name or Material 3-Hydroxy-2, 4, 5-trifluorobenzoic acid, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC357300250 SDS Thermo Fisher Scientific
AC357300250
Glass bottle 25g Each for $784.38
Specifications Description & Specifications

Chemical Identifiers

116751-24-7
100.0
C7 H3 F3 O3
192.09
3-Hydroxy-2, 4, 5-trifluorobenzoic acid, 98%
97.5
98%
F3C6H(OH)CO2H
MFCD00800378
≥97.5%

Specifications

Powder
25g
143° to 147°C
F3C6H(OH)CO2H
97.5
Authentic
C7 H3 F3 O3
3-Hydroxy-2, 4, 5-trifluorobenzoic acid, 98%
192.09
White to light yellow-beige
100.0
98%
Glass bottle
MFCD00800378
116751-24-7
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.