Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester

$133.80 - $483.68

Chemical Identifiers

Chemical Name or Material 3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester
CAS 1115-20-4
Molecular Formula C10H20O4
Formula Weight 204.27
MDL Number MFCD00059597
Synonym 3-Hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropionate; Hydroxypivalic acid neopentyl glycol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2596322

  SDS
alfa aesar™
H2596322
100g Each for $133.80

AAH2596336

  SDS
alfa aesar™
H2596336
500g Each for $483.68
Description & Specifications

Specifications

Formula Weight 204.27
CAS 1115-20-4
Molecular Formula C10H20O4
Chemical Name or Material 3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
Flash Point 161°C (322°F)
Melting Point 51° to 54°C
MDL Number MFCD00059597
Quantity 100g
Synonym 3-Hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropionate; Hydroxypivalic acid neopentyl glycol ester
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
TSCA TSCA
Boiling Point 292°C

Applications
3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester is a reactant in the Tischenko Reaction.

Solubility
Soluble in methanol(almost transparency).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.