Learn More
3-Hydroxy-1-methylpiperidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1082906
Description
Reactant for Optimization of Novobiocin scaffold to produce antitumor agents, Formation of carbamates and N-alkylimidazoles. Reactant for synthesis of Pyridine derivatives as CDK5 inhibitors Phenylcarbamate derivatives as ligands for nicotinic acetylcholine receptors, Amino phosphite ligands, Mexiletine enantiomers by nucleophilic aromatic substitution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant for Optimization of Novobiocin scaffold to produce antitumor agents, Formation of carbamates and N-alkylimidazoles. Reactant for synthesis of Pyridine derivatives as CDK5 inhibitors Phenylcarbamate derivatives as ligands for nicotinic acetylcholine receptors, Amino phosphite ligands, Mexiletine enantiomers by nucleophilic aromatic substitution.
Solubility
Fully misicible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
3-Hydroxy-1-methylpiperidine | |
0.999 | |
70°C (158°F) | |
1.474 | |
5 g | |
3-hydroxy-1-methylpiperidine, 1-methyl-3-piperidinol, n-methyl-3-piperidinol, 3-piperidinol, 1-methyl, 3-hydroxy-n-methylpiperidine, 1-methyl-3-hydroxypiperidine, n-methyl-3-hydroxypiperidine, n-methyl-3-hydroxy piperidine, 3r-1-methyl-3-piperidinol, pubchem6759 | |
UKANCZCEGQDKGF-UHFFFAOYSA-N | |
1-methylpiperidin-3-ol | |
98016 | |
98% |
3554-74-3 | |
76°C to 78°C (11 mmHg) | |
C6H13NO | |
MFCD00006495 | |
103017 | |
Fully misicible in water. | |
CN1CCCC(C1)O | |
115.176 | |
115.18 |
Chemical Identifiers
3554-74-3 | |
115.176 | |
UKANCZCEGQDKGF-UHFFFAOYSA-N | |
98016 | |
CN1CCCC(C1)O |
C6H13NO | |
MFCD00006495 | |
3-hydroxy-1-methylpiperidine, 1-methyl-3-piperidinol, n-methyl-3-piperidinol, 3-piperidinol, 1-methyl, 3-hydroxy-n-methylpiperidine, 1-methyl-3-hydroxypiperidine, n-methyl-3-hydroxypiperidine, n-methyl-3-hydroxy piperidine, 3r-1-methyl-3-piperidinol, pubchem6759 | |
1-methylpiperidin-3-ol |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 222-609-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only