missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxy-1-methacryloyloxyadamantane, TCI America™

$250.56 - $869.33

Chemical Identifiers

CAS 115372-36-6
Molecular Formula C14H20O3
Molecular Weight (g/mol) 236.311
MDL Number MFCD13152044
InChI Key OOIBFPKQHULHSQ-UHFFFAOYSA-N
Synonym 3-Hydroxy-1-adamantyl Methacrylate, Methacrylic Acid 3-Hydroxy-1-adamantyl Ester
PubChem CID 11856523
IUPAC Name (3-hydroxy-1-adamantyl) 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC12CC3CC(C1)CC(C3)(C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H154325G View Documents TCI America
H154325G
25g Each for $869.33
 
H15435G View Documents TCI America
H15435G
5g Each for $250.56
 
Specifications

Chemical Identifiers

115372-36-6
236.311
OOIBFPKQHULHSQ-UHFFFAOYSA-N
11856523
CC(=C)C(=O)OC12CC3CC(C1)CC(C3)(C2)O
C14H20O3
MFCD13152044
3-Hydroxy-1-adamantyl Methacrylate, Methacrylic Acid 3-Hydroxy-1-adamantyl Ester
(3-hydroxy-1-adamantyl) 2-methylprop-2-enoate

Specifications

White
25g
C14H20O3
3-Hydroxy-1-adamantyl Methacrylate, Methacrylic Acid 3-Hydroxy-1-adamantyl Ester
CC(=C)C(=O)OC12CC3CC(C1)CC(C3)(C2)O
236.311
236.31
≥98.0% (GC)
91°C
115372-36-6
MFCD13152044
OOIBFPKQHULHSQ-UHFFFAOYSA-N
(3-hydroxy-1-adamantyl) 2-methylprop-2-enoate
11856523
Crystal-Powder at 20°C
3-Hydroxy-1-methacryloyloxyadamantane
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1878

TSCA : Yes