missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Hydrazinocarbonyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H53365MD

 View more versions of this product

Catalog No. AAH53365MD



Specifications

Specifications

3-(Hydrazinocarbonyl)benzeneboronic acid
250mg
C7H9BN2O3
3-boronobenzohydrazide, 3-hydrazinecarbonyl benzeneboronic acid, benzoicacid, 3-borono-, hydrazide 9ci, 3-hydrazinecarbonyl phenylboronic acid, acmc-209ra2, 3-aminocarbamoylphenylboronic acid, benzoicacid,3-borono-,hydrazide 9ci, 3-hydrazinecarbonyl benzeneboronic aci, 3-hydrazinecarbonyl phenyl boronic acid
B(C1=CC(=CC=C1)C(=O)NN)(O)O
179.97
179.97
149°C to 151°C
913835-79-7
MFCD08689505
NKQNSAXAGLKZBP-UHFFFAOYSA-N
[3-(hydrazinecarbonyl)phenyl]boronic acid
44119369
97%

Chemical Identifiers

913835-79-7
179.97
NKQNSAXAGLKZBP-UHFFFAOYSA-N
44119369
B(C1=CC(=CC=C1)C(=O)NN)(O)O
C7H9BN2O3
MFCD08689505
3-boronobenzohydrazide, 3-hydrazinecarbonyl benzeneboronic acid, benzoicacid, 3-borono-, hydrazide 9ci, 3-hydrazinecarbonyl phenylboronic acid, acmc-209ra2, 3-aminocarbamoylphenylboronic acid, benzoicacid,3-borono-,hydrazide 9ci, 3-hydrazinecarbonyl benzeneboronic aci, 3-hydrazinecarbonyl phenyl boronic acid
[3-(hydrazinecarbonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents