Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hydrazinobenzoic acid, 99%

$77.59 - $1,129.84

Chemical Identifiers

Chemical Name or Material 3-Hydrazinobenzoic acid
CAS 38235-71-1
Molecular Formula C7H8N2O2
Formula Weight 152.15
MDL Number MFCD00191447
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2146209

 
alfa aesar™
B2146209
10g Each for $77.59

AAB2146218

 
alfa aesar™
B2146218
50g Each for $273.99

AAB2146230

 
alfa aesar™
B2146230
250g Each for $1,129.84
Description & Specifications

Specifications

Formula Weight 152.15
Chemical Name or Material 3-Hydrazinobenzoic acid
Assay 99%
Molecular Formula C7H8N2O2
MDL Number MFCD00191447
CAS 38235-71-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈194°C (dec.)
Quantity 10g

Applications
It finds its application oin the structure-activity studies of a series of dipyrazolo [3, 4-b: 3′, 4′-d] pyridin-3-ones binding to the immune regulatory protein B7. 1. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuffs.

Solubility
Very soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.