missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hexyne, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC308120050

 View more versions of this product

Catalog No. AC308120050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Liquid
Clear colorless to pale yellow
100.0
1.4100 to 1.4120 (20°C, 589nm)
Authentic
MFCD00009381
3-Hexyne
C6H10
82.15
5g
0.72g/mL
98.5
Glass bottle
CH3CHCCCHCH3
928-49-4
99%
99%

Chemical Identifiers

928-49-4
100.0
C6H10
82.15
3-Hexyne, 99%
98.5
99%
CH3CHCCCHCH3
MFCD00009381
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-173-4

TSCA : TSCA