Please login to your online account to display your discounted pricing

3-Hexyn-2-one, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Hexyn-2-one
CAS 1679-36-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H8O
Linear Formula CH3CH2Cidentical toCCOCH3
Formula Weight 96.13
MDL Number MFCD00041627
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379700050

  SDS
thermo fisher scientific
AC379700050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 96.13
Quantity 5g
Color Clear yellow
Density 0.8780g/mL
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Linear Formula CH3CH2Cidentical toCCOCH3
Infrared Spectrum Authentic
MDL Number MFCD00041627
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Hexyn-2-one
CAS 1679-36-3
Assay Percent Range 97%
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C6H8O
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Fatal if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Fatal if inhaled.
Flammable liquid and vapor.
Highly flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Fatal if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Fatal if inhaled.
Flammable liquid and vapor.
Highly flammable liquid and vapor.