Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hexyn-2-ol, 97%

Chemical Identifiers

Chemical Name or Material 3-Hexyn-2-ol
CAS 109-50-2
Molecular Formula C6H10O
Formula Weight 98.15
MDL Number MFCD00041586
UN Number UN1987
Beilstein 1739390
Synonym 1-Butynyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1157606

  SDS
alfa aesar™
L1157606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1157614

  SDS
alfa aesar™
L1157614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.15
Chemical Name or Material 3-Hexyn-2-ol
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C6H10O
Assay 97%
CAS 109-50-2
MDL Number MFCD00041586
Quantity 5g
Health Hazard 1 Warning
Synonym 1-Butynyl methyl carbinol
UN Number UN1987
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Beilstein 1739390
TSCA TSCA
Flash Point 54°C (129°F)
Boiling Point 79° to 80°C (60mmHg)
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.