missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hexyn-2-ol, 97%

Manufacturer:  Alfa Aesar™ L1157614

 View more versions of this product

Catalog No. AAL1157614



Specifications

Specifications

3-Hexyn-2-ol
79°C to 80°C (60mmHg)
109-50-2
MFCD00041586
1739390
IFCAMPHNVKBSTF-UHFFFAOYSA-N
hex-3-yn-2-ol
7990
97%
54°C (129°F)
25g
C6H10O
UN1987
3-hexyn-2-ol, hexyne-2-ol, acmc-20an2x, 3-hexyn-2-ol, s-2-benzyloxycarbonyl-4-methoxy-4-oxobutanoic acid
CCC#CC(C)O
98.145
98.15

Chemical Identifiers

109-50-2
98.145
IFCAMPHNVKBSTF-UHFFFAOYSA-N
7990
CCC#CC(C)O
C6H10O
MFCD00041586
3-hexyn-2-ol, hexyne-2-ol, acmc-20an2x, 3-hexyn-2-ol, s-2-benzyloxycarbonyl-4-methoxy-4-oxobutanoic acid
hex-3-yn-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

TSCA : TSCA

Documents