Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Hexanone, 97+%, Alfa Aesar™

$42.47 - $42.47

Chemical Identifiers

Chemical Name or Material 3-Hexanone
CAS 589-38-8
Molecular Formula C6H12O
Formula Weight 100.16
MDL Number MFCD00009402
UN Number UN1224
Beilstein 1738025
Synonym Ethyl n-propyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2079109

  SDS
alfa aesar™
B2079109
10g Each for $42.47

AAB2079118

  SDS
alfa aesar™
B2079118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2079130

  SDS
alfa aesar™
B2079130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.16
Chemical Name or Material 3-Hexanone
TSCA TSCA
Assay ≥97%
Molecular Formula C6H12O
MDL Number MFCD00009402
CAS 589-38-8
UN Number UN1224
Beilstein 1738025
Synonym Ethyl n-propyl ketone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Store in a well-ventilated place.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
RTECS Number MP1575000
Flash Point 15°C (59°F)
Odor Sweet
Boiling Point 119° to 121°C
Melting Point −55°C
Quantity 10g

Applications
3-Hexanone is a perfuming agent. It is used as a solvent. 3-Hexanone was used in a study to determine Henry′s law coefficients by combination of the equilibrium partitioning in closed systems technique in combination with solid-phase microextraction. It was used as test species in a study for quantitative determination of volatile organic compounds by chemical ionization reaction mass spectrometry. It was used to study the inhibitory activity of some aromatic and aliphatic ketones against Clostridium Botulinum spores and cells.

Solubility
Soluble in water. (14 g/L)

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Store in a well-ventilated place.

GHS H Statement
H225
Highly flammable liquid and vapour.