missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Heptyne, 97%

Manufacturer:  Alfa Aesar™ H5350414

 View more versions of this product

Catalog No. AAH5350414



Description

Description

Specifications

Specifications

3-Heptyne
−16°C (−3°F)
105°C to 106°C
2586-89-2
CH3(CH2)2C≡CCH2CH3
UN3295
1-ethyl-2-propylacetylene, 3-heptyne, acmc-20an2z
CCCC#CCC
96.173
96.17
0.741
1.4225
25g
C7H12
MFCD00039960
1731759
KLYHSJRCIZOUHE-UHFFFAOYSA-N
hept-3-yne
75755
97%

Chemical Identifiers

2586-89-2
96.173
KLYHSJRCIZOUHE-UHFFFAOYSA-N
75755
CCCC#CCC
C7H12
MFCD00039960
1-ethyl-2-propylacetylene, 3-heptyne, acmc-20an2z
hept-3-yne
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 219-967-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents