missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Heptylthiophene, TCI America™

$140.56 - $362.09

Chemical Identifiers

CAS 65016-61-7
Molecular Formula C11H18S
Molecular Weight (g/mol) 182.325
MDL Number MFCD00130131
InChI Key IUUMHORDQCAXQU-UHFFFAOYSA-N
Synonym 3-n-heptylthiophene, thiophene, 3-heptyl, unii-27b6l18uyc, acmc-209now, iuumhordqcaxqu-uhfffaoysa
PubChem CID 566850
IUPAC Name 3-heptylthiophene
SMILES CCCCCCCC1=CSC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H07221G View Documents TCI America
H07221G
1g Each for $140.56
 
H07225G View Documents TCI America
H07225G
5g Each for $362.09
 
Specifications

Chemical Identifiers

65016-61-7
182.325
IUUMHORDQCAXQU-UHFFFAOYSA-N
566850
CCCCCCCC1=CSC=C1
C11H18S
MFCD00130131
3-n-heptylthiophene, thiophene, 3-heptyl, unii-27b6l18uyc, acmc-209now, iuumhordqcaxqu-uhfffaoysa
3-heptylthiophene

Specifications

140°C
1g
C11H18S
3-n-heptylthiophene, thiophene, 3-heptyl, unii-27b6l18uyc, acmc-209now, iuumhordqcaxqu-uhfffaoysa
CCCCCCCC1=CSC=C1
182.325
182.33
≥94.0% (GC)
Yellow
65016-61-7
MFCD00130131
IUUMHORDQCAXQU-UHFFFAOYSA-N
3-heptylthiophene
566850
Clear Liquid at 20°C
3-Heptylthiophene
Safety and Handling

Safety and Handling

TSCA : No