missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Heptylpyrrole, TCI America™

$349.88 - $1,203.91

Chemical Identifiers

CAS 878-11-5
Molecular Formula C11H19N
Molecular Weight (g/mol) 165.28
MDL Number MFCD00191444
InChI Key OVBAZQHUSHSARW-UHFFFAOYSA-N
PubChem CID 258374
IUPAC Name 3-heptyl-1H-pyrrole
SMILES CCCCCCCC1=CNC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08095G View Documents TCI America
H08095G
5g Each for $1,203.91
 
H08091G View Documents TCI America
H08091G
1g Each for $349.88
 
Specifications

Chemical Identifiers

878-11-5
165.28
OVBAZQHUSHSARW-UHFFFAOYSA-N
3-heptyl-1H-pyrrole
C11H19N
MFCD00191444
258374
CCCCCCCC1=CNC=C1

Specifications

97°C
5g
C11H19N
OVBAZQHUSHSARW-UHFFFAOYSA-N
3-heptyl-1H-pyrrole
258374
Clear Liquid at 20°C
3-Heptylpyrrole
Brown
878-11-5
MFCD00191444
CCCCCCCC1=CNC=C1
165.28
165.28
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No