missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Heptanone, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 120380010
Specifications
3-Heptanone | |
41°C | |
Glass bottle | |
0.81 | |
Yellow | |
1L | |
98% | |
CH3(CH2)3COC2H5 | |
3-heptanone, aethylbutylketon, butyl ethyl ketone, eptan-3-one, ethyl butyl ketone, ethyl n-butyl ketone, ethylbutylcetone, ethylbutylketon, etilbutilchetone, n-butyl ethyl ketone | |
NGAZZOYFWWSOGK-UHFFFAOYSA-N | |
heptan-3-one | |
7802 | |
114.19 | |
98% |
0.8100g/mL | |
Authentic | |
1.4075 to 1.4095 | |
146.0°C to 149.0°C | |
-39.0°C | |
106-35-4 | |
C7H14O | |
01,699 | |
Solubility in water: 3.3g/L water (20° c). Other solubilities: soluble in alcohol and ether | |
CCCCC(=O)CC | |
114.19 | |
CHEBI:50139 | |
Liquid |
Chemical Identifiers
106-35-4 | |
114.19 | |
3-heptanone, aethylbutylketon, butyl ethyl ketone, eptan-3-one, ethyl butyl ketone, ethyl n-butyl ketone, ethylbutylcetone, ethylbutylketon, etilbutilchetone, n-butyl ethyl ketone | |
CHEBI:50139 | |
CCCCC(=O)CC |
C7H14O | |
NGAZZOYFWWSOGK-UHFFFAOYSA-N | |
7802 | |
heptan-3-one |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 203-388-1