missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Guanidinopropionic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 426660050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Guanidinopropionic acid | |
97% | |
C4H9N3O2 | |
MFCD00045939 | |
3-guanidinopropionic acid, 3-guanidinopropanoic acid, beta-guanidinopropionic acid, guanidinopropionic acid, 3-guanidinopropanoate, 3-carbamimidamidopropanoic acid, beta-gpa, unii-ul1984yrka, beta-guanadinopropionate, n-aminoiminomethyl-beta-alanine | |
C(CN=C(N)N)C(=O)O | |
131.13 | |
CHEBI:15968 | |
97% |
353-09-3 | |
Glass bottle | |
H2NC(=NH)NH(CH2)2CO2H | |
5g | |
KMXXSJLYVJEBHI-UHFFFAOYSA-N | |
3-(diaminomethylideneamino)propanoic acid | |
67701 | |
131.13 |
Chemical Identifiers
353-09-3 | |
131.13 | |
KMXXSJLYVJEBHI-UHFFFAOYSA-N | |
67701 | |
3-(diaminomethylideneamino)propanoic acid |
C4H9N3O2 | |
MFCD00045939 | |
3-guanidinopropionic acid, 3-guanidinopropanoic acid, beta-guanidinopropionic acid, guanidinopropionic acid, 3-guanidinopropanoate, 3-carbamimidamidopropanoic acid, beta-gpa, unii-ul1984yrka, beta-guanadinopropionate, n-aminoiminomethyl-beta-alanine | |
CHEBI:15968 | |
C(CN=C(N)N)C(=O)O |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 206-530-
RUO â Research Use Only