Please login to your online account to display your discounted pricing

3-Glycidoxypropyltrimethoxysilane, 97%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  216545000

 View more versions of this product

Catalog No. AC216545000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (GC)
Physical Form Liquid
Formula Weight 236.34
Quantity 500g
Color Clear
Density 1.0700g/mL
Beilstein 17, V,3, 45
Chemical Name or Material 3-Glycidoxypropyltrimethoxysilane
Packaging Glass bottle
Synonym GLYMO
CAS 2530-83-8
Assay Percent Range 97%
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
Health Hazard 2 GHS P Statement Causes serious eye damage.
Health Hazard 1 Danger
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement Wear face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C9H20O5Si

Specifications

Chemical Name or Material 3-Glycidoxypropyltrimethoxysilane
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C9H20O5Si
Formula Weight 236.34
Synonym GLYMO
Accessories

Danger

GHS P Statement Wear face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Causes serious eye damage.

SDS & Safety Documents