missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Glycidoxypropyltrimethoxysilane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 216541000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Glycidoxypropyltrimethoxysilane | |
1.0700g/mL | |
122°C | |
96% min. (GC) | |
C9H20O5Si | |
100 g | |
1.07 | |
Solubility in water: soluble. Other solubilities: soluble in acetone, benzene and ether | |
CO[Si](CCCOCC1CO1)(OC)OC | |
236.34 | |
4 mPa.s (20°C) | |
97% |
2530-83-8 | |
120.0°C (2.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4280 to 1.4300 | |
17, V,3, 45 | |
3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane | |
BPSIOYPQMFLKFR-UHFFFAOYSA-N | |
trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane | |
17317 | |
236.34 | |
Liquid |
Chemical Identifiers
2530-83-8 | |
236.34 | |
3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane | |
trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane |
C9H20O5Si | |
BPSIOYPQMFLKFR-UHFFFAOYSA-N | |
17317 | |
CO[Si](CCCOCC1CO1)(OC)OC |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 219-784-2
RUO – Research Use Only