Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Furonitrile, 98%

Chemical Identifiers

Chemical Name or Material 3-Furonitrile
CAS 30078-65-0
Molecular Formula C5H3NO
Formula Weight 93.09
MDL Number MFCD00067042
UN Number UN2926
Beilstein 507009
Synonym 3-Cyanofuran; Furan-3-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1401903

  SDS
alfa aesar™
L1401903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL14019MD

  SDS
alfa aesar™
L14019MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1401906

  SDS
alfa aesar™
L1401906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 93.09
Beilstein 507009
Flash Point 53°C (127°F)
Chemical Name or Material 3-Furonitrile
Assay 98%
CAS 30078-65-0
MDL Number MFCD00067042
Health Hazard 3 GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 150° to 151°C
Melting Point 28° to 30°C
Synonym 3-Cyanofuran; Furan-3-carbonitrile
UN Number UN2926
Health Hazard 2 GHS H Statement
H331-H228-H302-H312
Toxic if inhaled.
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Molecular Formula C5H3NO
Health Hazard 1 Danger
Quantity 1g
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H228-H302-H312
Toxic if inhaled.
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.