Please login to your online account to display your discounted pricing

3-Furoic acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Furoic acid
Name Note 99%
CAS 488-93-7
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C5H4O3
Formula Weight 112.08
MDL Number MFCD00005350
Synonym 3-Furancarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC172450050

  SDS
thermo fisher scientific
172450050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.08
Quantity 5g
EINECS Number 207-689-9
TSCA TSCA
Beilstein 18, I, 439
CAS Max % 100.0
CAS Min % 98.5
Packaging Glass bottle
Synonym 3-Furancarboxylic acid
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C5H4O3
Name Note 99%
CAS 488-93-7
Chemical Name or Material 3-Furoic acid
Assay Percent Range 99%
MDL Number MFCD00005350
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.