Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester, 97%

$427.92 - $427.92

Chemical Identifiers

Chemical Name or Material 3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester
CAS 1073353-63-5
Molecular Formula C18H22BNO4
Formula Weight 327.18
MDL Number MFCD09266173
Synonym 3-(Furfurylaminocarbonyl)phenylboronic acid pinacol ester; N-Furfuryl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2859906

 
alfa aesar™
H2859906
5g Each for $427.92
Description & Specifications

Specifications

Formula Weight 327.18
Chemical Name or Material 3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C18H22BNO4
MDL Number MFCD09266173
CAS 1073353-63-5
Synonym 3-(Furfurylaminocarbonyl)phenylboronic acid pinacol ester; N-Furfuryl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Melting Point 122° to 124°C
Quantity 5g