Please login to your online account to display your discounted pricing

3-Furanmethanol, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Furanmethanol
CAS 4412-91-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H6O2
Formula Weight 98.1
MDL Number MFCD00005352
Synonym 3-Hydroxymethylfuran
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC188080050

  SDS
thermo fisher scientific
AC188080050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC188080250

  SDS
thermo fisher scientific
AC188080250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5%
Formula Weight 98.1
Quantity 5g
Color Clear colorless to slightly yellow
Beilstein 17, III, 1266
Density 1.1390g/mL
CAS Max % 100.0
Chemical Name or Material 3-Furanmethanol
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C5H6O2
Refractive Index 1.4835 to 1.4855 (20°C, 589nm)
MDL Number MFCD00005352
CAS 4412-91-3
Synonym 3-Hydroxymethylfuran
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.