missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Furancarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America F03045G
Specifications
3-Furancarboxylic Acid | |
122°C | |
MFCD00005350 | |
3-furoic acid, 3-furancarboxylic acid, 3-carboxyfuran, 3-furanoic acid, unii-88a3s7rg98, 3-furoate, 3-furans, 3-furanoate, pubchem6945, acmc-209kee | |
OC(=O)C1=COC=C1 | |
112.08 | |
CHEBI:30846 | |
≥98.0% (T) |
488-93-7 | |
C5H4O3 | |
5 g | |
IHCCAYCGZOLTEU-UHFFFAOYSA-N | |
furan-3-carboxylic acid | |
10268 | |
112.08 | |
Crystalline Powder |
Chemical Identifiers
488-93-7 | |
112.08 | |
IHCCAYCGZOLTEU-UHFFFAOYSA-N | |
10268 | |
furan-3-carboxylic acid |
C5H4O3 | |
MFCD00005350 | |
3-furoic acid, 3-furancarboxylic acid, 3-carboxyfuran, 3-furanoic acid, unii-88a3s7rg98, 3-furoate, 3-furans, 3-furanoate, pubchem6945, acmc-209kee | |
CHEBI:30846 | |
OC(=O)C1=COC=C1 |
Safety and Handling
TSCA : No