Please login to your online account to display your discounted pricing

3-Furancarboximidamide hydrochloride, Technical Grade, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Furancarboximidamide hydrochloride
Assay Tech.
Molecular Formula C5H6N2O·ClH
Formula Weight 146.58
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00101CB

 
fisher scientific company
MO00101CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Formula Weight 146.58
Quantity 250mg
Assay Tech.
Chemical Name or Material 3-Furancarboximidamide hydrochloride
Molecular Formula C5H6N2O·ClH
CAS 54649-21-7