Please login to your online account to display your discounted pricing

3-Furancarboximidamide hydrochloride, Technical Grade, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO00101CB

 View more versions of this product

Catalog No. MO00101CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 146.58
Purity Grade Technical Grade
Quantity 250mg
Assay Tech.
CAS 54649-21-7
Molecular Formula C5H6N2O·ClH
Chemical Name or Material 3-Furancarboximidamide hydrochloride

Specifications

Chemical Name or Material 3-Furancarboximidamide hydrochloride
Assay Tech.
Molecular Formula C5H6N2O·ClH
Formula Weight 146.58