Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Furaldehyde, 97%, stab. with ≈0.05% BHT

Chemical Identifiers

Chemical Name or Material 3-Furaldehyde
CAS 498-60-2
Name Note Stabilized with ≈0.05% BHT
Molecular Formula C5H4O2
Formula Weight 96.09
MDL Number MFCD00010424
UN Number UN1199
Beilstein 105852
Synonym Furan-3-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1133406

  SDS
alfa aesar™
A1133406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1133414

  SDS
alfa aesar™
A1133414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1133422

  SDS
alfa aesar™
A1133422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 96.09
UN Number UN1199
Assay 97%
CAS 498-60-2
Flash Point 48°C (118°F)
Chemical Name or Material 3-Furaldehyde
Boiling Point 145°C
Recommended Storage 2°C to 8°C
Health Hazard 1 Danger
Beilstein 105852
Odor Almond-like
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: FURALDEHYDES
Molecular Formula C5H4O2
Synonym Furan-3-carboxaldehyde
Quantity 5g
Health Hazard 2 GHS H Statement
H300-H330-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note Stabilized with ≈0.05% BHT
MDL Number MFCD00010424

Applications
3-Furaldehyde is compound of honey of various floral origin. It is used to produce N-furan-3-ylmethylene-N′-pyridin-2-yl-hydrazine by reaction with 2-hydrazino-pyridine. It is also used as a solvent.

Solubility
Soluble in water, benzene, chloroform, alcohol and ether.

Notes
Air & Light Sensitive. Store at 4°C. Store in the dark. Incompatible with bases, light and air.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2°C to 8°C