missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Formylphenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC359370050

 View more versions of this product

Catalog No. AC359370050



Description

Description

Specifications

Specifications

3-Formylphenylboronic acid
Glass bottle
100°C
87199-16-4
100.0
C7H7BO3
3-boronobenzaldehyde, 3-dihydroxyboryl benzaldehyde, 3-formyl phenylboronic acid, 3-formylbenzeneboronic acid, 3-formylboronic acid, 3-formylphenyl boronic acid, 3-formylphenylboronicacid, boronic acid, 3-formylphenyl, m-formylphenylboronic acid
OB(O)C1=CC=CC(C=O)=C1
149.94
149.94
97%
Authentic
Beige to White or Orange
5g
96.0
96% min. (HPLC)
MFCD00161356
HJBGZJMKTOMQRR-UHFFFAOYSA-N
(3-formylphenyl)boronic acid
2734356
Powder, Crystals, and/or Chunks

Chemical Identifiers

87199-16-4
149.94
HJBGZJMKTOMQRR-UHFFFAOYSA-N
2734356
OB(O)C1=CC=CC(C=O)=C1
C7H7BO3
MFCD00161356
3-boronobenzaldehyde, 3-dihydroxyboryl benzaldehyde, 3-formyl phenylboronic acid, 3-formylbenzeneboronic acid, 3-formylboronic acid, 3-formylphenyl boronic acid, 3-formylphenylboronicacid, boronic acid, 3-formylphenyl, m-formylphenylboronic acid
(3-formylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents