Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Formylindole-1-acetic acid, 97+%

$77.27 - $301.70

Chemical Identifiers

Chemical Name or Material 3-Formylindole-1-acetic acid
CAS 138423-98-0
Molecular Formula C11H9NO3
Formula Weight 203.2
MDL Number MFCD00452923
Beilstein 5431204
Synonym 1-(Carboxymethyl)indole-3-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2680703

 
alfa aesar™
H2680703
1g Each for $77.27

AAH2680706

 
alfa aesar™
H2680706
5g Each for $301.70
Description & Specifications

Specifications

Formula Weight 203.2
Molecular Formula C11H9NO3
Beilstein 5431204
Melting Point ≈205°C
Chemical Name or Material 3-Formylindole-1-acetic acid
Assay ≥97%
MDL Number MFCD00452923
CAS 138423-98-0
Synonym 1-(Carboxymethyl)indole-3-carboxaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.