Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Formylfuran-2-boronic acid, 97%

$136.69 - $532.59

Chemical Identifiers

Chemical Name or Material 3-Formylfuran-2-boronic acid
CAS 27339-38-4
Molecular Formula C5H5BO4
Formula Weight 139.9
MDL Number MFCD01075680
Beilstein 1637137
Synonym 2-Borono-3-furaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1519803

  SDS
alfa aesar™
L1519803
1g Each for $136.69

AAL1519806

  SDS
alfa aesar™
L1519806
5g Each for $532.59
Description & Specifications

Specifications

Formula Weight 139.9
Assay 97%
Molecular Formula C5H5BO4
MDL Number MFCD01075680
CAS 27339-38-4
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈150°C
Chemical Name or Material 3-Formylfuran-2-boronic acid
Beilstein 1637137
Synonym 2-Borono-3-furaldehyde
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.