missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Formyl-6-methylchromone, 97%, ACROS Organics™

Chemical Identifiers

CAS 42059-81-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H8O3
Formula Weight 188.18
MDL Number MFCD00138943
Synonym 6-Methyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde
Chemical Name or Material 3-Formyl-6-methylchromone
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC370140250 SDS Thermo Fisher Scientific
AC370140250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

42059-81-4
100.0
C11H8O3
MFCD00138943
3-Formyl-6-methylchromone
96.0
97%
188.18
6-Methyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde
≥96.0%

Specifications

Yellow
25g
96.0
Authentic
Powder
C11H8O3
MFCD00138943
3-Formyl-6-methylchromone
169° to 174°C
42059-81-4
100.0
Glass bottle
97%
188.18
6-Methyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde
≥96.0%