Please login to your online account to display your discounted pricing

3'-Formyl(1,1'-biphenyl)-4-carboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO01256DA

 View more versions of this product

Catalog No. MO01256DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 226.23
Quantity 1g
Assay 95%
CAS Min % 95.0
Molecular Formula C14H10O3
MDL Number MFCD04039069
CAS 222180-23-6
Synonym 3«-Formyl(1, 1«-biphenyl)-4-carboxylic acid (IUPAC)
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3'-Formyl(1, 1'-biphenyl)-4-carboxylic acid
Packaging Amber Glass Bottle
Assay Percent Range 95%

Specifications

Chemical Name or Material 3'-Formyl(1, 1'-biphenyl)-4-carboxylic acid
CAS 222180-23-6
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C14H10O3
Formula Weight 226.23
MDL Number MFCD04039069
Synonym 3«-Formyl(1, 1«-biphenyl)-4-carboxylic acid (IUPAC)
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents