Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Fmoc-aminomethyl)pyridine, 97%

Chemical Identifiers

Chemical Name or Material 3-(Fmoc-aminomethyl)pyridine
Formula Weight 330.38
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6687503

 
alfa aesar™
H6687503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6687506

 
alfa aesar™
H6687506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 330.38
Chemical Name or Material 3-(Fmoc-aminomethyl)pyridine
Physical Form Solid
Assay 97%
Color White
Quantity 1g