missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorotoluene, TCI America™

Manufacturer:  TCI America F0040500ML

 View more versions of this product

Catalog No. F0040500ML



Specifications

Specifications

3-Fluorotoluene
Yellow
352-70-5
MFCD00000339
3-fluorotoluene, m-fluorotoluene, benzene, 1-fluoro-3-methyl, 1-fluoro-3-methyl-benzene, 1-methyl-3-fluorobenzene, toluene, m-fluoro, unii-f72w445dq0, 3-fluorobenzyl radical, meta-fluorotoluene, acmc-209ifn
CC1=CC(=CC=C1)F
110.131
110.13
≥99.0% (GC)
115°C
500mL
C7H7F
2388
BTQZKHUEUDPRST-UHFFFAOYSA-N
1-fluoro-3-methylbenzene
9606
Clear Liquid at 20°C

Chemical Identifiers

352-70-5
110.131
BTQZKHUEUDPRST-UHFFFAOYSA-N
9606
CC1=CC(=CC=C1)F
C7H7F
MFCD00000339
3-fluorotoluene, m-fluorotoluene, benzene, 1-fluoro-3-methyl, 1-fluoro-3-methyl-benzene, 1-methyl-3-fluorobenzene, toluene, m-fluoro, unii-f72w445dq0, 3-fluorobenzyl radical, meta-fluorotoluene, acmc-209ifn
1-fluoro-3-methylbenzene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0083

TSCA : Yes

Documents