Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol

Chemical Identifiers

Chemical Name or Material 3-Fluorostyrene
CAS 350-51-6
Name Note Stabilized with 0.1% 4-tert-butylcatechol
Molecular Formula C8H7F
Formula Weight 122.14
MDL Number MFCD00000341
UN Number UN1993
Beilstein 2038488
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1240603

 
alfa aesar™
A1240603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1240606

 
alfa aesar™
A1240606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.14
Chemical Name or Material 3-Fluorostyrene
Name Note Stabilized with 0.1% 4-tert-butylcatechol
Assay 97%
Molecular Formula C8H7F
MDL Number MFCD00000341
CAS 350-51-6
UN Number UN1993
Beilstein 2038488
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2°C to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 29°C (84°F)
Odor Recognizable
Boiling Point 40° to 42°C (8mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2°C to 8°C