Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorosalicylic acid, 99%

$129.27 - $323.96

Chemical Identifiers

Chemical Name or Material 3-Fluorosalicylic acid
CAS 341-27-5
Molecular Formula C7H5FO3
Formula Weight 156.11
MDL Number MFCD00153167
Synonym 3-Fluoro-2-hydroxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2616304

  SDS
alfa aesar™
H2616304
2g Each for $323.96

AAH26163ME

  SDS
alfa aesar™
H26163ME
500mg Each for $129.27
Description & Specifications

Specifications

Formula Weight 156.11
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 2g
Chemical Name or Material 3-Fluorosalicylic acid
Assay 99%
Molecular Formula C7H5FO3
MDL Number MFCD00153167
CAS 341-27-5
Synonym 3-Fluoro-2-hydroxybenzoic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 146° to 151°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.