Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoropyridine N-oxide, 98%

$124.68 - $504.88

Chemical Identifiers

Chemical Name or Material 3-Fluoropyridine N-oxide
CAS 695-37-4
Molecular Formula C5H4FNO
Formula Weight 113.09
MDL Number MFCD03095181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2576506

 
alfa aesar™
H2576506
5g Each for $124.68

AAH2576514

 
alfa aesar™
H2576514
25g Each for $504.88
Description & Specifications

Specifications

Formula Weight 113.09
Chemical Name or Material 3-Fluoropyridine N-oxide
Assay 98%
Molecular Formula C5H4FNO
MDL Number MFCD03095181
CAS 695-37-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 65° to 68°C
Quantity 5g

Applications
3-Fluoropyridine N-oxide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.