Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoropyridine-4-boronic acid pinacol ester, 97%

$88.65 - $304.78

Chemical Identifiers

Chemical Name or Material 3-Fluoropyridine-4-boronic acid pinacol ester
CAS 458532-88-2
Molecular Formula C11H15BFNO2
Formula Weight 223.06
MDL Number MFCD06798251
Synonym 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5006903

 
alfa aesar™
H5006903
1g Each for $304.78

AAH50069MD

 
alfa aesar™
H50069MD
250mg Each for $88.65
Description & Specifications

Specifications

Formula Weight 223.06
Chemical Name or Material 3-Fluoropyridine-4-boronic acid pinacol ester
Assay 97%
Molecular Formula C11H15BFNO2
MDL Number MFCD06798251
CAS 458532-88-2
Synonym 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 125° to 127°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.